Anti-VIM Mouse Monoclonal Antibody (Biotin) [clone: VM1170]
Supplier: Biotium
BNCB1170-500 BNCB1170-100
BNCB1170-500EA 560 EUR
BNCB1170-500 BNCB1170-100
Anti-VIM Mouse Monoclonal Antibody (Biotin) [clone: VM1170]
Antibodies
This MAb reacts with a 58 kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP s) such as desmin, keratin, neurofilament, and glial fibrillary acid protein.Anti-vimentin alone is of limited value as a diagnostic tool; however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain vimentin, e.g. hepatoma and seminoma. Anti-vimentin is also useful as a tissue process control reagent.
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: VM1170
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human