Anti-HSP90B1 Mouse Monoclonal Antibody (CF405S) [clone: HSP90B1/1192]

Supplier: Biotium
BNC041192-500 BNC041192-100
BTIUBNC041192-500EA 560 EUR
BTIUBNC041192-500 BTIUBNC041192-100
Anti-HSP90B1 Mouse Monoclonal Antibody (CF405S) [clone: HSP90B1/1192]
Antibodies
This antibody recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: HSP90B1/1192
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol HSP90B1
Antigen name Glucose Regulated Protein 94
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone Clone HSP90B1/1192
Reactivity Human
Host Mouse
Gene ID 192374
Isotype IgG2a kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR