Anti-UACA Mouse Monoclonal Antibody (CF594) [clone: UACA/1222]
Supplier: Biotium
BNC941222-500 BNC941222-100
BNC941222-500EA 560 EUR
BNC941222-500 BNC941222-100
Anti-UACA Mouse Monoclonal Antibody (CF594) [clone: UACA/1222]
Antibodies
UACA (Uveal Autoantigen with Coiled-coil domains and Ankyrin repeats) is a 1,416 amino acid nuclear membrane protein. It was originally identified as an autoantigen in patients with panuveitis, a characteristic of Vogt-Koyanagi-Harada disease, and in patients with Graves' disease. UACA was also later identified as Nucling, an mRNA differentially expressed in F9 embryonal carcinoma cells during cardiac muscle differentiation. UACA appears to function as a pro-apoptotic protein that recruits the apaf-1-pro-caspase-9 complex for the induction of apoptosis to mediate the cell-death pathway.
Type: Primary
Antigen: UACA
Clonality: Monoclonal
Clone: UACA/1222
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse