Anti-NAPSA Mouse Monoclonal Antibody (Biotin) [clone: NAPSA/1239]

Supplier: Biotium
BNCB1239-500 BNCB1239-100
BNCB1239-500EA 560 EUR
BNCB1239-500 BNCB1239-100
Anti-NAPSA Mouse Monoclonal Antibody (Biotin) [clone: NAPSA/1239]
Antibodies
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.

Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: NAPSA/1239
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR