Capillary columns, MXT®-624
Supplier: Restek
70968 70969 70971 70973
REST70968EA 942 EUR
REST70968 REST70969 REST70971 REST70973
Capillary columns, MXT®-624
Chromatography Columns
MXT®-624 Siltek®-treated stainless steel columns are application specific columns for volatile organic pollutants. Recommended in U.S. EPA methods for volatile organic pollutants.
  • Low- to midpolarity phase
  • Temperature range form –20 to 280 °C
  • Equivalent to USP G43 phase
  • 4,5 inch standard coil diameter