Sulphuric acid 95,85 - 96,15% 96% calibration solution of viscosity acc. to DIN EN ISO 307
Supplier: VWR Chemicals
Danger
85652.290EA 96.6 EUR
85652.290 85652.320
Sulphuric acid 95,85 - 96,15% 96% calibration solution of viscosity acc. to DIN EN ISO 307
Sulphuric acid
Delivery information: This product is not available in all countries. Please check with your local VWR office or supplier.
Formula: H₂SO₄
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 10.38 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
REACH: 01-2119458838-20
Merck Index: 14,08974
Order Now
Specification Test Results
Assay 95.85 - 96.15 %