MXT®-union connector kits for fused silica columns
Supplier: Restek
21384 21385 21386 21387 21388 21389
REST21384EA 222 EUR
REST21384 REST21385 REST21386 REST21387 REST21388 REST21389
MXT®-union connector kits for fused silica columns
Fittings and Connectors
These MXT® union connectors kits are suitable for connecting metal and/or fused silica GC columns.
  • Reusable, low dead volume and leak-tight connection
  • Siltek treatment ensures maximum inertness
  • Use to oven temperatures of 350 °C
  • Available in union and 'Y' configurations.
These MXT union connectors can be used with fused silica tubing. This unique graphite-reinforced composite allows a capillary column to slide into the adaptor and be locked in place simply by loosening and tightening the fitting.
Delivery information: Each MXT®-union connector kit is supplied with the MXT® union, two 1/32 inch nuts and two one-piece fused silica adaptors. Each MXT® 'Y'-union connector kit includes the MXT® union, three 1/32 inch nuts and three one-piece fused silica adaptors.