NGAL (pig) ELISA kit
Supplier: Enzo Life Sciences
ALEXBPD-KIT-044EA 1410 EUR
ALEXBPD-KIT-044 ENZOBPDKIT044
NGAL (pig) ELISA kit
Assays ELISAs
For the quantitative determination of NGAL in pig urine, plasma, serum, tissue extracts, or culture supernatants.
  • Ready to use pre-coated plate and individual calibrators save time and reduce errors
  • Easy to use with convenient storage at 4 °C and room temperature incubations
  • Absorbance is read at 450 nm
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in pig samples. Absorbance is read at 450nm.
Delivery information: The kit contains a microwell plate coated with anti-NGAL (pig) antibody (96 wells), 5X sample diluent concentration, pig NGAL calibrator (1-7. 0, 10, 20, 40, 100, 200, 400 pg/ml), 25X wash solution concentration, biotinylated pig-NGAL antibody, HRP-streptavidin, TMB substrate and 12 ml stop solution.