VWR® EML Digital Plus Test Sieve Shakers
Supplier: NEXOPART
510-0957EA 3700 EUR
510-0957 510-0954 510-0963 510-0960 510-0959 510-0956 510-0965 510-0962 510-1291 510-1297 510-1300 510-1289 510-1298 510-1295 510-1294 510-1292 HAVE205322730 HAVE205322747 HAVE205322754 HAVE205322860 HAVE205322877 HAVE205322884 HAVE205322891
VWR® EML Digital Plus Test Sieve Shakers
Shakers Sieve Shakers
These electromagnetically-driven test sieve shakers are designed for sieve analysis with optimum three-dimensional sieving action. The self-re-adjusting amplitude, which uses permanent acceleration measurement of the whole sieve tower, balances the oscillation of the shaker and its stand. This guarantees that amplitude stays the same, irrespective of the feed quantity and the installation position.
  • Memory for 10 test sieve parameter sets
  • Timer function: 0 to 99 minutes or continuous operation
  • Intermittent or continuous shaking operation
  • Digital display of all functions
  • 3000 impulses per minute (50 Hz)
  • Sound emission: 70 dB(A) for EML 315 digital plus, EML 450 digital plus: 73 dB(A)
  • Maximum loading: 21 kg for EML 315 (6 kg sieved material), 42 kg for EML 450 (15 kg sieved material)
EML 315 and 450 models are for dry and wet sieving and have a separate control device.
Optional HAVER 'TwinNut' the quick-release clamping system:
'TwinNut' clamps for easy fixing and removal of the cover are an alternative to standard 'Classic' knurled nuts. The guide rods of the 'Classic' system can also be used for the HAVER 'TwinNut' system.
Delivery information: Included with each shaker: Models with either 'Classic' clamping system or 'TwinNut' system are supplied with two guide rods (660 mm for EML 200 and EML 315; 850 or 1000 mm for EML 450) and two knurled nuts with handlebars, cover with inspection glass and water spray diffuser for wet sieving models. Sieves and collecting pan must be ordered separately. IQ/OQ documentation on request.