High-capacity moisture traps
Supplier: Restek
21997 20638
REST21997EA 224 EUR
REST21997 REST20638
High-capacity moisture traps
Filters Gas Filters
These high-capacity moisture traps are purged with ultra-high-purity helium and are ready to be used with any carrier gas including hydrogen.
  • Reduces water to less than 15 ppb
  • Moisture capacity: 16 g of water
Maximum operating pressure: 250 psi
Maximum flow: 1,25 l/min