Low-dead-volume connector kits, MXT®
Supplier: Restek
20394 20395 20396 20397
REST20394EA 174 EUR
REST20394 REST20395 REST20396 REST20397
Low-dead-volume connector kits, MXT®
Fittings and Connectors
Rugged MXT® low-dead-volume connectors are Siltek® treated to make them inert to active compounds. They can be used at temperatures up to 430 °C without degrading the deactivated layer.
  • Ideal for metal columns
  • Stainless steel ferrules and nuts
  • Low thermal mass tracks rapid oven temperature programming
Standard connector kit connects a guard column/transfer line to an MXT stainless steel column. 'Y' connector kit connects two MXT columns to one inlet or one MXT column to two detectors.
Delivery information: Each kit contains the MXT union, two (standard kit) or three ('Y' connector kit) 0,8 mm ferrules and nuts.