Capillary column nuts, finger-tight
Supplier: Restek
21040 21041
REST21040EA 69 EUR
REST21040 REST21041
Capillary column nuts, finger-tight
Fittings and Connectors
These finger-tight capillary column nuts made from high-quality stainless steel are an alternative to conventional column nuts. These fittings work best with graphite type ferrules and help prevent over tightening, which can cause graphite material to exude into the inlet.
  • Allow wrench-free column installations
  • Available for standard (1,58 mm) and compact (Agilent-style) ferrules
These fittings can be installed by hand without a wrench and cool down much faster than conventional fittings, making column and inlet maintenance easier.