Capillary columns, MXT®-65 (Siltek®-treated stainless steel)
Supplier: Restek
77020 77023 77035 77038 77050 77053
REST77020EA 0 EUR
REST77020 REST77023 REST77035 REST77038 REST77050 REST77053
Capillary columns, MXT®-65 (Siltek®-treated stainless steel)
Chromatography Columns
MXT®-65 columns are general-purpose columns for phenols, triglycerides, and fatty acids.
  • Mid to high polarity phase
  • Crossbond® diphenyl dimethyl polysiloxane
  • Temperature range: 50 to 300 °C
  • Equivalent to USP G17 phase