Gas-tight syringes, PTFE tipped (SGE)
Supplier: Restek
21220 24789 24797
REST21220EA 152 EUR
REST21220 REST24789 REST24797
Gas-tight syringes, PTFE tipped (SGE)
Syringes Syringes for Chromatography
These tips are used for Agilent autosamplers 7673, 7683, 7693A, and 6850.