Thermal gas purifiers
Supplier: Restek
21496 21497 21498 21499
REST21496EA 0 EUR
REST21496 REST21497 REST21498 REST21499
Thermal gas purifiers
Filters Gas Filters
This thermal gas purifier removes oxygen, water, carbon monoxide, carbon dioxide, hydrocarbons (except methane) to ppb levels which is pure enough for MS.
  • Packed with reactor-grade, pure getter material for maximum efficiency and no contamination
  • Welded end-fittings on getter tubes for leak free connection
  • Each tube has 12 l oxygen and 35 l water vapor capacity
  • With the max. flow rate of 1 L/min
The getter material in line of thermal gas purifiers reacts chemically with impurities in the carrier gas stream. Because the reaction is non-reversible, there is no possibility of contaminants breaking through the thermal gas purifier.