VWR® Flasks, Flat Bottom, with Standard Ground Joint
Supplier: VWR Collection
201-1381EA 19.1 EUR
201-1381 201-1380 201-1382 201-1383 201-1384 201-1344 201-1348 201-1360 201-1361
VWR® Flasks, Flat Bottom, with Standard Ground Joint
Flasks Flat Bottom Flasks
Borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Inexpensive alternative for a wide range of laboratory applications. With white labelling area and imprinted catalogue number for convenient reordering.
DIN 12348
Certifications: DIN 12348