VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
201-1351EA 14.8 EUR
201-1351 201-1352 201-1353 201-1354 201-1355 201-1356 201-1357 201-1358 201-1359 201-1321 201-1322 201-1326 201-1342 201-1343
VWR® Flasks, Round Bottom, with Standard Ground joint
Flasks Round Bottom Flasks
Borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Inexpensive alternative for a wide range of laboratory applications. With white labelling area and imprinted catalogue number for convenient reordering.
DIN 12348
Certifications: DIN 12348