Bottle-top dispensers and titrators, opus®
Supplier: HIRSCHMANN
9581020 9581050 9582020 9582050 9581010 9582010
613-0755EA 2366 EUR
613-0755 613-0756 613-0761 613-0762 613-1594 613-1596
Bottle-top dispensers and titrators, opus®
Bottle Top Dispensers
Safe and reliable, these bottle-top dispensers and titrators provide extremely sensitive, motor-powered dispensing precision. These devices operate with remote touch screen controllers that give complete control over all functions.
  • Controllers feature menu-guided user instructions with tab cards on a remote, coloured TFT touch screen
  • Stepper function enables you to dispense several volume increments from a single cylinder fill
  • Pumps equipped with ceramic compound piston seals for excellent durability and electronic protection. A return valve at the base of the pump helps reduce costs by enabling users to prime the pump and return liquids to the bottle
  • Optional silicone system platform neatly organises all components by securely combining bottle and pump and providing a convenient space for controller placement
  • Other options: Battery pack to operate independently from any power outlet, language selection (English, German, French and Spanish), specification of suction and dispensing speed and the capacity for up to ten different memory settings further enhance the versatility of the controllers
PC interface: RS232 or USB
Accuracy: 0,6% - dispenser 10/20/50 ml; 0,5% - titrator 20/50 ml
Precision (CV): 0,2% - dispenser 10/20/50 ml; 0,07/0,07/0,05% - titrator 10/20/50 ml
Ordering information: Supplied as complete kits with bottle-top mounted pump, free-standing display and individual certificate of calibration.