VWR® Forceps
Supplier: VWR Collection
232-0150EA 35.9 EUR
232-0150 232-0151 232-0152 232-0153 232-0154 232-0155
VWR® Forceps
Forceps
PTFE, white.
  • Very good chemical resistance, inert, non stick
  • Temperature resistant from –200 to +280 °C
  • Smooth surfaces prevent contamination and allow easy cleaning