384-well (HTS) microplates, non treated, BRANDplates®
Supplier: Brand
781620 781621 781622 781626 781627 781680 781681 781682 781686 781687
735-2010EA 175 EUR
735-2010 735-2011 735-2012 735-2016 735-2017 735-2030 735-2031 735-2032 735-2036 735-2037
384-well (HTS) microplates, non treated, BRANDplates®
Microplates HTS Microplates
These PS, flat bottom plates are available solid black, white or transparent.
  • All BRAND® plates are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analyses
  • Sterile plates are supplied individually wrapped with lid
pureGrade™: Non treated, medium binding surface, non sterile. These plates are suitable for homogeneous assays, screening and storage.
pureGrade™ S: Non treated, sterile. Sterilised by β radiation, these plates are especially well suited for bacteriological assays.
Non treated, medium binding, sterile surface. Sterilised by β radiation, these plates are especially well suited for bacteriological assays.
Certifications: BRANDplates® are non cytotoxic according to ISO 10993-5, free of endotoxins (<0,01 EU/ml), and free from DNA, DNase and RNase. Sterile products comply with ISO 11137 and AAMI guidelines, SAL 10-⁶.