Selfpacker® Preparative HPLC Columns
Supplier: MERKLINGER PRAZISION WERKZEUG
9250161016 9250161018 9250161019 1250050001 9250161017 1259600001 9250162702 9250162701 9250162700 9250161020 1259120001 1259350001 1259340001 1259320001 1259310001 1259300001 1259400001 1259110001 1259090001 1259080001 1250140001 1250130001 1250100001 1259550001 1259530001 1259510001 1259500001 1259480001 1259380001 1259460001 1259430001 1259420001 1259410001 1259610001 1259490001 9250163009 9250164707 9250165017 9250163025 9250163024 9250163023 9250163022 9250163021 9250164710 9250163029 9250163016 9250163020 9250164706 9250164705 9250164704 9250164703 9250164702 9250163026 9250163027 9250163030 9250164708 G8005.115 G8005.125 1259130001 1259140001 1259370001
556-0032EA 285 EUR
556-0032 556-0034 556-0035 556-0000 556-0033 556-0028 556-0039 556-0038 556-0037 556-0036 556-0007 556-0014 556-0013 556-0012 556-0011 556-0010 556-0017 556-0006 556-0005 556-0004 556-0003 556-0002 556-0001 556-0027 556-0026 556-0025 556-0024 556-0022 556-0016 556-0021 556-0020 556-0019 556-0018 556-0029 556-0023 556-0040 556-0057 556-0060 556-0047 556-0046 556-0045 556-0044 556-0043 556-0059 556-0050 556-0041 556-0042 556-0056 556-0055 556-0054 556-0053 556-0052 556-0048 556-0049 556-0051 556-0058 556-0030 556-0031 556-0072 556-0073 556-0074
Selfpacker® Preparative HPLC Columns
Chromatography Columns
This column-packing technology is based on a combination of vacuum suction with subsequent column bed compression that stabilises the packing. A common water-jet pump is used to generate the vacuum. The sorbent slurry is sucked through a packing reservoir into the column by applying low pressure at the bottom end of the column.
  • Allows precise adjustment of the desired packing volume or sorbent bed height
  • Facilitates the determination of the column geometry
  • Makes direct scale-up from analytical to preparative scale easy
Packing stand NW25
Min./max. packing height: 185/220 mm
Max. working pressure: 200 bar
Packing stand NW50
Min./max. packing height: 185/220 mm
Max. working pressure: 100 bar
Packing stand NW100
Min./max. packing height: 295/350 mm
Max. working pressure: 100 bar
Max. working pressure: 200 bar