Industrial gloves, oil repellent, HyFlex®
Supplier: Ansell
11-948/10 11-948/7 11-948/8 11-948/9 11-947/7 11-947/8 11-947/9 11-947/10 11-947/11 11-948/11
ANSE11-948/10EA 276 EUR
ANSE11-948/10 ANSE11-948/7 ANSE11-948/8 ANSE11-948/9 ANSE11-947/7 ANSE11-947/8 ANSE11-947/9 ANSE11-947/10 ANSE11-947/11 ANSE11-948/11
Industrial gloves, oil repellent, HyFlex®
Gloves
These seamless knitted liners with Intercept™ technology yarn either feature a ¾ dipped or a full nitrile coating.
  • Excellent abrasion performance
  • Good blade cut resistance
  • Very good grip properties due to the nitrile micro finishing of the coating
  • Flexible and comfortable
  • Coating resistant to residual oils and greases present on handled parts
Intercept™: Specialised knitting technology which blends engineered, synthetic and natural fibers into high performance yarns. This provides high-cut protection with exceptional comfort and dexterity.
Fortix®More Product Information
Order Now
SPECIFICATIONS
Model 11-948 11-947 11-948
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific Hand-specific Hand-specific
Disposable / reusable Reusable Reusable Reusable
Latex / latex-free Latex
Liner material Intercept™ yarn Intercept™ yarn Intercept™ yarn
Finish ¾ dipped ¾ dipped ¾ dipped
Gauge 13 13 13
Cuff style Safety cuff Knit wrist Safety cuff
EN norm EN 388:2016 (4X42B) EN 388:2016 (4X42B) EN 388:2016 (4X42B)