Lubrol, white waxy solid

Supplier: MP Biomedicals

Synonyms: Polyethylene Glycol Monohexadecyl Ether, Polyoxyethylene monoacetyl ether, Polyethylene glycolmonocetyl ether

0219529980 0219529983
ICNA0219529980EA 99.4 EUR
ICNA0219529980 ICNA0219529983
Lubrol, white waxy solid
Lubrol

Lubrol is a compound composed of ethylene oxide condensates of fatty alcohols. Lubrol is used as a solubilizing agent for membrane bound proteins and membrane-bound adenylcyclase. This material is a non-ionic surfactant, emulsifier.


Form: Lubrol WX. This material is an ethylene oxide condensate of fatty alcohols; cetyl-stearyl ether
pH (5% aqueous solution): 7.0 - 8.5
Solubility: Soluble in water (5% w/v-clear, colorless solution)

Formula: HO(CH₂CH₂O)nC₁₆H₃₃ n= ~23
Storage Temperature: Ambient
MDL Number: MFCD00080892
CAS Number: 9004-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR