Percoll®, yellow liquid

Supplier: MP Biomedicals

0219536925 0219536980 0219536990
ICNA0219536925EA 78.7 EUR
ICNA0219536925 ICNA0219536980 ICNA0219536990
Percoll®, yellow liquid
Percoll®

Colloidal PVP coated silica for cell separation. Aseptically filled.


Storage temperature.: +4 °C


pH: 8.9 ± 0.3 at 20°C (lit.)

Storage Temperature: Refrigerator
MDL Number: MFCD00131830
CAS Number: 65455-52-9

Order Now

Specification Test Results

Identity Test Passes
Viscosity Record cp at 20 deg C
Osmolality Max 25 mOsm/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR