Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Ultrapure
Supplier: MP Biomedicals
Warning
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
04819620 04819623 04819638
ICNA04819620EA 132 EUR
ICNA04819620 ICNA04819623 ICNA04819638
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Ultrapure
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Biological buffer reagent.
  • Synonyms: Tromethamine
  • Heavy metals (as Pb): <5 ppm
  • O.D. (260nm) of a 5% saturated solution: <0.1
  • pKa at 20°C = 8.1
pH: As Tris solutions decrease in temperature from 25°C to 5°C, the pH value increases an average of 0.03 units per°C. As the solution increases in temperature from 25°C to 37°C, the pH decreases an average of 0.025 units per°C.
Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM