Reservoir system
Supplier: Beckman Coulter
372790 372795 372784
BECL372790EA 278 EUR
BECL372790 BECL372795 BECL372784
Reservoir system
Reagent Reservoirs
This modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling systems including the full suite of Biomek liquid handlers.
  • Half and quarter reservoirs must be used with a frame
  • Full reservoirs hold up to 150 ml of liquid, and do not require a frame
  • Each frame can accommodate up to four quarter reservoirs or two half reservoirs
  • All reservoirs are sterile and made of PP
Not for use with 96 and 384 multi channel pipetting heads.