Safety helmets, uvex pheos E-WR
Supplier: UVEX
9770.030 9770.130 9770.230 9770.530 9770.330 9770.430
111-2337EA 19.4 EUR
111-2337 111-2338 111-2339 111-2342 111-2340 111-2341
Safety helmets, uvex pheos E-WR
Head Protection Safety Helmets
The lightweight uvex pheos safety helmets feature a sporty design and offer maximum wearer comfort. The sturdy, rigid helmet shell also easily withstands pressure from the side, and fulfills all safety-related requirements for construction or industrial work.
  • Can quickly and easily be adapted to the user's individual needs - visor, safety spectacles, ear defenders or head torch
  • Suspension harness with wheel ratchet for variable width adjustment
  • Six-point textile band attachment ensures optimal fit and comfort
  • Climazone ventilation and long brim
  • With slot on the front for helmet lights and side Euroslot adapters (30 mm) for attaching earmuffs and the uvex pheos visor system
Suitable for use by electricians as helmet shell is fully enclosed.
Size: 520 to 610 mm
Certifications: EN 397 and requirements for very low temperatures (–30 °C) and molten metal (MM) splash; EN 50365 (1000 VAC).