J.T.Baker® BAKERBOND spe™ Anhydrous Sodium Sulfate-Florisil, SPE Columns for Special Applications
Supplier: Avantor
7495-04EA 366.94 EUR
7495-04 7495-18
J.T.Baker® BAKERBOND spe™ Anhydrous Sodium Sulfate-Florisil, SPE Columns for Special Applications
Chromatography Columns
These SPE columns are designed for the clean-up of extracts of hydrocarbons from water.
Certifications: ISO 9377-2