Cation exchange chromatography columns, TSK-GEL® SP-5PW
Supplier: TOSOH Bioscience
0007161 0007211 0007575 0008803 0008807 0013062 0014017 0016093 0018758
TOYO0007161EA 1950 EUR
TOYO0007161 TOYO0007211 TOYO0007575 TOYO0008803 TOYO0008807 TOYO0013062 TOYO0014017 TOYO0016093 TOYO0018758
Cation exchange chromatography columns, TSK-GEL® SP-5PW
Chromatography Columns
TSK-GEL® SP-5PW are polymer based column packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. Used for the separation and analysis of proteins, peptides, and other biologically active molecules.