J.T.Baker® BAKERBOND® BDC C18, HPLC Columns
Supplier: Avantor
7441-00EA 1215.14 EUR
7441-00 7441-01
J.T.Baker® BAKERBOND® BDC C18, HPLC Columns
Chromatography Columns
These columns are ideal for the determination of pharmaceutical active components. They are endcapped and suitable for chromatography of basic compounds.
  • Excellent column to column and batch to batch reproduciblity
  • Sharp peaks and improved symmetry
The BAKERBOND® BDC C18 phase receives a special base deactivation treatment during silica material manufacture to reduce silanophilic activity.