Full face masks, X-plore® 5500
Supplier: Dräger
R55270 R56655
111-0417EA 152 EUR
111-0417 111-0408
Full face masks, X-plore® 5500
Respirators Reusable Respirators Full Face Respirators
X-plore® 5500 full face masks offer the perfect solution wherever your work requires respiratory protection, e.g. in the chemical industry, metalworking and automotive industries, shipbuilding, maintenance, supply and disposal logistics. The double layered face seal with triple sealing action provides reliable protection and a tight fit for all face shapes and sizes. The large visor is available in two different materials: polycarbonate or triplex (excellent anti-scratch coating and high chemical-resistance).
  • The filters are low-profile and positioned on the side, which makes for a wide field of vision and excellent weight distribution for extensive freedom of movement
  • Two bayonet connections to enable filters to be fitted simply and safely
  • The high-quality clear visor lens provides a wide, extensive field of vision and high chemical, thermal and scratch resistance
  • Five-point head strap makes the mask quick and easy to fasten without trapping hair
Weight: 540 g
Certifications: EN 136
Caution: *One size fits all.