Slim Jim® Vented Waste Bin Accessories
Supplier: Rubbermaid Commercial Products
FG268888LGRAY FG270388BLUE FG269288GRN FG267360GRAY FG267400BLA FG269288RED FG267360BLUE FG267360BLA FG269288YEL 1968468 1788372 1788373 1956191 FG269288BLUE 1980602
129-3701EA 69.8 EUR
129-3701 129-3703 129-0443 129-0327 129-0328 129-0329 129-0442 129-0420 129-0423 129-2028 129-2174 129-2175 129-2089 129-1874 RUBB1980602
Slim Jim® Vented Waste Bin Accessories
Bins Waste Bins
Transform the Slim Jim® Vented container in combination with the dollies into a mobile station or with the various lid options into a small recycling station.
The accessories are compatible with the Slim Jim® containers 60 and 87 L (not compatible with the Slim Jim® Recycling station).
The Slim Jim® lids from Rubbermaid Commercial make waste separation easier with interchangeable, colour-coded tops. The entire Slim Jim® lid range consists of:
• Lid with slot opening (blue) for paper waste.
• Lid with round opening (green or yellow) for glass bottles and cans.
• Lid with propeller opening (green or blue) for mixed waste.
• Swing lid (black, blue, or grey), hinged lid (black), or lid with handle (grey) for general waste.
• 3 YEAR WARRANTY comes with a 3 year warranty against normal wear and tear, our commitment to guaranteed durability.
The Slim Jim® dollies from Rubbermaid Commercial are designed to transport Slim Jim® containers effortlessly and efficiently through all commercial areas.
Slim Jim® Dollies (Plastic):
• For efficient collection and transport, can combine multiple dollies via a side connection.
• The dollies can be easily and quickly separated using a foot pedal.
• Structural foam construction ensures maximum durability in commercial environments.
• 3 YEAR WARRANTY comes with a 3 year warranty against normal wear and tear; our commitment to guaranteed durability.
Slim Jim® Dollies (Stainless steel):
• Durable Construction: Made from high-quality stainless steel, which is easy to clean and does not rust or chip in damp environments.
• Quiet Maneuverability: With abrasion-resistant and quiet 7,6 cm wheels that allow smooth and silent maneuverability.
• Lockable Wheels: The locking function of the wheels ensures a stable recycling station or collection point.
• Durable Wheels: Includes swivel casters made of plastic that are resistant to chemicals, rust, and corrosion even in wet environments.
• 3 YEAR WARRANTY comes with a 3 year warranty against normal wear and tear; our commitment to guaranteed durability.