Cold resistant gloves, ActivArmr® 23-700
Supplier: Ansell
23-700/10 23-700/9
112-0270EA 29.6 EUR
112-0270 112-2926
Cold resistant gloves, ActivArmr® 23-700
Gloves
These fully coated gloves feature a PU foam insulation for additional warmth and comfort. The textured surface provides unbeatable grip on wet and slippery surfaces. The robust, liquid-repellent PVC coating does not absorb water, does not stiffen like leather and remains flexible even in sub zero temperatures.
  • Salt, chemical and corrosion resistant - does not crack or split
  • Ergonomic fit for increased comfort - reduces finger and hand fatigue
  • Water-repellent, turn-out lining carries sweat away from hand
Ideal for food processing and automotive applications.
Certifications: EN 388:2016 (3121B), EN 511 (111), EN ISO 374-1 Type B (KLT), EN ISO 374-5; Cat. III