Chemical resistant gloves, FineDex® 944-32 Fisherman
Supplier: HONEYWELL SAFETY
2094432/7 2094432/8 2094432/9 2094432/10
112-0161EA 109 EUR
112-0161 112-0164 112-0170 112-0192
Chemical resistant gloves, FineDex® 944-32 Fisherman
Gloves
These 100% polyamide knitted liners feature a latex coating which offers good mechanical and chemical resistance against detergents and liquid products. It also resists acids and bases. The glove's crepe finish helps to improve the grip on slippery parts (tailored for the fishing trades).
  • Natural latex provides excellent flexibility, dexterity and comfort
  • Knitted, seamless lining makes the glove ever so close to the hand
  • Elasticated at the wrist
  • Crinkled finishing: Ideal for fish handling
  • Extended cuff helps to protect a large part of the forearm
Ideal for use in fishing, fish processing, ice handling, sorting and packing, and wine growing.
Certifications: EN 388:2016 (3142X), EN ISO 374:2016 Type A (AKLMNO), EN 407 (X2XXXX); Cat. II
Food contact approved.