VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
612-4466EA 19.3 EUR
612-4466 612-4518 612-4468 612-4469 612-4470 612-4471 612-4472 612-4473 612-4520 612-4476 612-4479 612-4481 612-4486 612-4490 612-4491 612-4492 612-4494 612-4495 612-4496 612-4497 612-4499 612-4500 612-4502 612-4543 612-4504 612-4505 612-4506 612-4508 612-4509 612-4511 612-4512 612-4513 612-4514 612-4515 612-4542 612-4533 612-4526 612-4527 612-4530 612-4523 612-4536 612-4538 612-4545 612-3399 612-3608 612-3609 612-3610 612-3619 612-3627 612-5770 612-5781
VWR® Standard Line Transfer Pipettes
Pipettes Pasteur/Transfer Pipettes
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
  • Will not shatter or break
  • Non toxic and inert to biological fluids and most acids
  • Can be sealed, refrigerated and gas sterilised
  • No bulb to insert or remove
  • Uniform drop size
The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
Ordering information: To help you find the correct product and catalogue number, please have a look at the brochure VWR® for Liquid Handling, pages 12 to 13, which you can find under 'Related information' and 'Product literature'. There are images of the pipettes available next to the catalogue number and description of packaging.