ESD safety shoes, lace-up, A460
Supplier: ATLAS - the shoe company
78800_S1/39 78800_S1/36 78800_S1/37 78800_S1/38 78800_S1/40 78800_S1/41 78800_S1/42 78800_S1/43
114-7533EA 0 EUR
114-7533 114-7530 114-7531 114-7532 114-7534 114-7535 114-7536 114-7537
ESD safety shoes, lace-up, A460
Shoes and Boots
Sneaker style ESD safety shoes made from Sportline upper (oiled nubuck leather) and a Sneaker Line sole (3,0 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: Pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Protective steel toecap (200 J)
  • Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Blue
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1