ESD safety shoes, lace-up, SL 20 Green
Supplier: ATLAS - the shoe company
293/35 293/36 293/37 293/38 293/39 293/40 293/41 293/42 293/43 293/44 293/45 293/46 293/47 293/48 293/49
ATLA293/35EA 198 EUR
ATLA293/35 ATLA293/36 ATLA293/37 ATLA293/38 ATLA293/39 ATLA293/40 ATLA293/41 ATLA293/42 ATLA293/43 ATLA293/44 ATLA293/45 ATLA293/46 ATLA293/47 ATLA293/48 ATLA293/49
ESD safety shoes, lace-up, SL 20 Green
Shoes and Boots
Lightweight and metal-free ESD safety shoes made from Sportline upper (oiled nubuck leather), light lining materials and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Lightweight alu-tec® toecap made from aluminium
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black/green
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC