Imipenem monohydrate
Supplier: Cayman Chemical
Synonyms: (5R,6S)-3-((2-Formimidamidoethyl)thio)-6-((R)-1-hydroxyethyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate
16039-10 16039-25 16039-100 16039-5
CAYM16039-10EA 104 EUR
CAYM16039-10 CAYM16039-25 CAYM16039-100 CAYM16039-5
Imipenem monohydrate
Imipenem monohydrate
Imipenem is an intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0,5 - 2 µg/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species. Imipenem acts by interfering with bacterial cell wall synthesis.
  • A crystalline solid
Formula: C₁₂H₁₇N₃O₄S
MW: 317.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09753321
CAS Number: 74431-23-5