Potassium hydrogen phthalate ≥99.95% ACS

Supplier: Hach

HACH31534EA 166 EUR
HACH31534
Potassium hydrogen phthalate ≥99.95% ACS
Potassium hydrogen phthalate
Formula: HOOCC₆H₄COOK
MW: 204,22 g/mol
MDL Number: MFCD00013070
CAS Number: 877-24-7
https://ru.vwr.com/stibo/web/std.lang.all/06/58/16640658.jpg

Order Now

Specification Test Results

Assay 99,95 - 100,05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR