Specimen containers, Samco™ Clicktainer™
Supplier: SAMCO
90WHT53-2000 90YEL53-2000 90WHT53-2002 90WHT53-2003 90WHT53-2001 90WHT53-2004 90WHT53-1002 90WHT53-1001 90ORG53-1000 90WHT53-1000 90GRN53-1000 90ORG53-1001
216-2174EA 79.3 EUR
216-2174 216-2179 216-2176 216-2177 216-2175 216-2178 216-2173 216-2172 216-2169 216-2171 216-2168 216-2170
Specimen containers, Samco™ Clicktainer™
Sample Containers General Purpose Containers
These Samco™ Clicktainer™ vials have a wide opening and leakproof seal. The innovative 'click' seal closure system makes these vials well suited for the collection and transport of fluids, solids and powders.
  • Visual, audible and tactile indication of full cap closure (HDPE)
  • Safe tab design reduces risk of tear to gloves or break off during use
  • Wide 53 mm opening for ease of sample collection and management
  • 'Cick' seal system indicates when the cap is properly applied so the sample is secure
  • Ideal for use in hospital pneumatic transport systems
  • Excellent clarity with easy to see graduations
  • Available in both sterile and non-sterile versions
Ideal for a wide range of customer applications, from the collection of valuable and often hazardous patient samples such as urine, faeces and sputum and their transportation through hospital pneumatic / vacuum transport systems to their suitability for use with formalin or fixatives for histology applications.
Wide range of cap colours are available to help with sample differentiation and management. Vials are available as assembled product for general laboratory use, clinical applications and mice necropsy. Dispenser box is ideal for use both in smaller laboratories and doctors offices. Vials have clearly designed label for patient fields and required information.
Certifications: Each batch pressure tested to 95 kPa in accordance with BS EN 14401. Leak tested in accordance with BS EN 14254. CE marked in accordance with IVD Directicve 98/79/EC.