Fondaparinux Sodium, Reference Standards
Supplier: USP
1282707 1282718 1282730
1282707.EA 509 EUR
1282707. 1282718. 1282730.
Fondaparinux Sodium, Reference Standards
Standards Organic Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Fondaparinux Sodium System Suitability Mixture B: A mixture containing fondaparinux sodium and fondaparinux related compound A, B, C, D, E, F, and G
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required