Loperamide Related Compounds, Reference Standard

Supplier: USP

1370000 1370010 1370065
1370000.EA 385 EUR
1370000. 1370010. 1370065.
Loperamide Related Compounds, Reference Standard
Standards Organic Standards

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certifications: US Pharmacopea Reference Standard

Order Now

SPECIFICATIONS

Description Loperamide Hydrochloride Loperamide Loperamide Related Compound F
Synonym(s) 1r,4s)-4-(4-Chlorophenyl)-1-[4-(dimethylamino)-4-oxo-3,3-diphenylbutyl]-4-hydroxypiperidine 1-oxide hydrate
Environmentally Preferable
CAS No. 34552-83-5 106900-12-3
Formula C₂₉H₃₄Cl₂N₂O₂
UN No. 2811 2811 2811

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR