Metronidazole, Reference Standards
Supplier: USP
1442009 1442100
1442009.EA 385 EUR
1442009. 1442100.
Metronidazole, Reference Standards
Standards Organic Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Order Now
SPECIFICATIONS
Description Metronidazole Metronidazole Benzoate
Environmentally Preferable
CAS No. 443-48-1 13182-89-3
Formula C₆H₉N₃O₃ C₁₃H₁₃N₃O₄