Adenosine 5′-triphosphate magnesium salt ≥95%, Sigma-Aldrich®

Supplier: Merck
Warning

A9187-100MG A9187-500MG A9187-1G A9187-5X1G
SIALA9187-100MGEA 65 EUR
SIALA9187-100MG SIALA9187-500MG SIALA9187-1G SIALA9187-5X1G
Adenosine 5′-triphosphate magnesium salt ≥95%, Sigma-Aldrich®
Adenosine 5′-triphosphate magnesium salt

Adenosine 5γ-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is use in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP.


  • ATP provides the metabolic energy to drive metabolic pumps
  • It serves as a coenzyme in a wide array of enzymatic reactions


Biological source: bacterial

Formula: C₁₀H₁₆N₅O₁₃P₃·xMg²⁺
MDL Number: MFCD00084767
CAS Number: 74804-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR