Casein Kinase I substrate
Supplier: Merck
C2335-1MG
C2335-250UG
SIALC2335-1MGEA
721
EUR
SIALC2335-1MG
SIALC2335-250UG
Casein Kinase I substrate
Proteins and Peptides
Casein kinase-1 is 6-fold higher than Vmax in caesin.
Potency: 172 μM Km
Composition: peptide content, ~70%
Packaging: Packed in bottomless glass bottle and contents are inside inserted fused cone.