Papaverine hydrochloride ≥98%, white powder, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
P3510-5G
P3510-100G
P3510-25G
SIALP3510-5GEA
69
EUR
SIALP3510-5G
SIALP3510-100G
SIALP3510-25G
Papaverine hydrochloride ≥98%, white powder, Sigma-Aldrich®
Papaverine hydrochloride
Smooth muscle relaxant and cerebral vasodilator; phosphodiesterase inhibitor.
- White powder
- Solubility in water: 25 mg/ml
Formula:
C₂₀H₂₁NO₄·HCl |
MDL Number:
MFCD00012745 CAS Number: 61-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...