8-Quinolinesulphonyl chloride 98%
Supplier: Thermo Fisher Scientific
Danger
132210050 132210250
ACRO132210050EA 59.1 EUR
ACRO132210050 ACRO132210250
8-Quinolinesulphonyl chloride 98%
8-Quinolinesulphonyl chloride
Formula: C₉H₆ClNO₂S
Boiling Pt: 306 °C (760 mmHg)
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00006808
CAS Number: 18704-37-5
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
Titration Argentometric ≥97.5 %