Ammonium cerium(IV) nitrate, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate
212360010 212360050
ACRO212360010EA 248 EUR
ACRO212360010 ACRO212360050
Ammonium cerium(IV) nitrate, pure
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
UN: 3085
ADR: 5.1 II
Merck Index: 13,00509
Order Now
Specification Test Results
Appearance Orange Crystalline powder or crystals
Loss on drying ≤2 %
Titration ≥98.5 %