Trimethylsilanol
Supplier: Thermo Fisher Scientific
Danger
458570050 458570250 458571000
ACRO458570050EA 47.5 EUR
ACRO458570050 ACRO458570250 ACRO458571000
Trimethylsilanol
Trimethylsilanol
Formula: C₃H₁₀OSi
Boiling Pt: 98 °C
Melting Pt: –12 °C
Density: 0.814 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD02751657
CAS Number: 1066-40-6
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %