4-Azaphthalide 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-(Hydroxymethyl)nicotinic acid gamma-lactone, Furo[3,4-b]pyridin-5(7H)-one
458450010 458450050 458450250
ACRO458450010EA 94.6 EUR
ACRO458450010 ACRO458450050 ACRO458450250
4-Azaphthalide 98%
4-Azaphthalide
Formula: C₇H₅NO₂
MW: 135.12 g/mol
Melting Pt: 143…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01860224
CAS Number: 5657-51-2
Order Now
Specification Test Results
Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 140 to 146 °C
HPLC ≥97.5 %